2',5'-Didesoxyadenosin ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

BMLCN1100005 BMLCN1100025
ENZOBMLCN1100005EA 165 EUR
ENZOBMLCN1100005 ENZOBMLCN1100025
2',5'-Didesoxyadenosin ≥98% (durch TLC)
2',5'-Didesoxyadenosin

Adenylate cyclase inhibitor


A useful, cell-permeable inhibitor of adenylate cyclase (ID50=250µM). Inhibits growth and caues cell differentiation (1.5µM) in K562 cells.

Formel: C₁₀H₁₃N₅O₂
MDL: MFCD00210904
CAS-Nummer: 6698-26-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ab68638f143149a79ebd3bac821ea05e3418c80&vid=cc06f876d932bc8df02eac72521b6066164f9ad3&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR