Rp-cAMPS ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

BMLCN1350001
ENZOBMLCN1350001EA 161 EUR
ENZOBMLCN1350001
Rp-cAMPS ≥98% (durch HPLC)
Rp-cAMPS

Rp-cAMPS is a potent and specific competitive inhibitor of the activation of cAMP-dependent protein kinases by cAMP (IC50=4.9 µM) . Its cell permeability and complete resistance to cyclic nucleotide phosphodiesterases make it a unique tool for studying cAMP-dependent signaling. Significantly more lipophilc as compared to cAMP and thus membrane permeable in many biological systems. This product is not sterile.


  • PKA inhibitor


White solid.


Soluble in water (25mg/ml), aqueous buffers, methanol, 100% ethanol or DMSO.

Formel: C₁₀H₁₁N₅O₅PS·C₆H₁₆N
Molecular Weight: 446,5 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 151837-09-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=672f732e0d0c132c58a947681c9dc5c529d3c463&vid=4ae1175351feea7b510c528d02b9f02a2ff3fabe&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR