Sp-cAMPS ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

BMLCN1360001
ENZOBMLCN1360001EA 86.9 EUR
ENZOBMLCN1360001
Sp-cAMPS ≥98% (durch HPLC)
Sp-cAMPS

PKA activator.


Sp-cAMPS is a potent and specific activator of cAMP-dependent protein kinases. Its cell permeability and complete resistance to cyclic nucleotide phosphodiesterases make it a unique tool for studying cAMP-dependent signaling. Has a considerably higher resistance against cyclic nucleotide-dependent phosphodiesterases as compared to dibutyryl-cAMP or 8-Br-cAMP. Concentration for half-maximal activation is 1.8 µM. The product is not sterile. No metabolic side effects.


Soluble in water (25 mg/ml), DMSO, dimethyl formamide or methanol.

Formel: C₁₀H₁₁N₅O₅PS·C₆H₁₆N
Lagertemperatur: Gefrierschrank
CAS-Nummer: 93602-66-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd9c83d128b7937f07d7593834fc0b9822a834b1&vid=b245e1bbc9656b0d6bb575999106cc0c99b3b2dd&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Purity ≥98% (HPLC)
Identity Determined by UV-VIS and ESI-MS

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR