Rp-8-Br-cGMPS ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

BMLCN2160500
ENZOBMLCN2160500EA 208 EUR
ENZOBMLCN2160500
Rp-8-Br-cGMPS ≥98% (durch HPLC)
Rp-8-Br-cGMPS

Cell-permeable, phosphodiesterase-resistant cGMP antagonist. Reversed NO stimulation of random migration in rabbit neutrophils. Used at 30µM to cometively inhibit 8-Br-cGMP relaxation of rabbit aortas. The product is not sterile. Resistant to nucleotide-dependent phosphodiesterases; no metabolic side effects. Significantly more lipophilic and membrane permeant as compared to cyclic GMP or Rp-cGMPS.


  • PKG inhibitor


White lyophilized powder.


Soluble in water (25mg/ml).

Formel: C₁₀H₁₀BrN₅O₆PS·Na
Molecular Weight: 462,2 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 150418-07-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ea621688420dc61123e06c47b1d4e0a44acba75&vid=e543da520e3eecba871e82c721b6d3e5b838ea65&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR