Fumagillin ≥95% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

BMLCT1000005
ENZOBMLCT1000005EA 498 EUR
ENZOBMLCT1000005
Fumagillin ≥95% (durch TLC)
Fumagillin

Methionine aminopeptidase 2 inhibitor


Inhibitor of angiogenesis and endothelial cell proliferation. Specific inhibitor of methionine aminopeptidase type II (MetAP-II) by covalently modifying a conserved active-site histidine. Precursor to TNP-470. Exhibits antitumor activity in estrogen-induced pituitary adenoma. Antineoplastic. Anti-infective.


Isolated from Aspergillus fumigatus.

Formel: C₂₆H₃₄O₇
MDL: MFCD03990453
CAS-Nummer: 23110-15-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6e812745b76551ff3f756c5501642ed1bd43acc&vid=e82df31e482c19b5dd24d38c87c739280d32125c&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR