Cytochalasin E ≥98% (durch HPLC, TLC)
Lieferant: ENZO LIFE SCIENCES
BMLCT1200005
ENZOBMLCT1200005EA
299
EUR
ENZOBMLCT1200005
Cytochalasin E ≥98% (durch HPLC, TLC)
Cytochalasin E
Inhibits angiogenesis
An aspergillus derived fungal metabolite. Inhibits angiogenesis and tumor growth. An actin microfilament depolymerizing agent.
Formel:
C₂₈H₃₃NO₇ Lagertemperatur: Gefrierschrank |
CAS-Nummer:
36011-19-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...