1-Stearoyl-2-arachidonoyl-sn-glycerol ≥95% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

BMLDG1300010
ENZOBMLDG1300010EA 352 EUR
ENZOBMLDG1300010
1-Stearoyl-2-arachidonoyl-sn-glycerol ≥95% (durch TLC)
1-Stearoyl-2-arachidonoyl-sn-glycerol

PKC activator.


Putative endogenous activator of protein kinase C produced via receptor-mediated inositol phospholipid hydrolysis.


Soluble in DMSO or 100% ethanol.

Formel: C₄₁H₇₂O₅
Molecular Weight: 645,0 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 65914-84-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9184b232c29bd1f7753afdfc3e1fd7de38eaeac8&vid=49e6e77452204d9f0e5604958dbceb4427fd0df1&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Purity ≥95% (TLC)
Appearance Oil or waxy solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR