25-Hydroxyvitamin D₂ ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES
Gefahr

Synonyme: 25-Hydroxyergocalciferol, 25-Hydroxyvitamin D2, Ercalcidiol, 25-Hydroxycalciferol, (S,Z)-3-(2-((1R,3aS,7aR,E)-1-((2R,5S,E)-6-Hydroxy-5,6-dimethylhept-3-en-2-yl)-7a-methyloctahydro-4H-inden-4-ylidene)ethylidene)-4-methylenecyclohexan-1-ol

BMLDM1010001
ENZOBMLDM1010001EA 228 EUR
ENZOBMLDM1010001
25-Hydroxyvitamin D₂ ≥98% (durch HPLC)
25-Hydroxyvitamin D₂

High purity vitamin D₂ intermediate.


25-Hydroxyvitamin D₂ is an intermediate produced from vitamin D₂ by cytochrome P450 isoforms in serum. This primary metabolite of vitamin D₂ can be used to indirectly measure the concencentration of vitamin D via HPLC and UV.


Soluble in DMSO, ethanol or chloroform.

Formel: C₂₈H₄₄O₂
Molecular Weight: 412,65 g/mol
MDL: MFCD02683431
CAS-Nummer: 21343-40-8
UN: 2811
ADR: 6.1,II

Bestellen

Spezifikation Testergebnisse

Purity (HPLC) ≥98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR