5(S),6(R)-Dihydroxyeicosa-7E,9E,11Z,14Z-tetraenoic acid (DiHETE) ≥99% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

BMLDS0560050
ENZOBMLDS0560050EA 270 EUR
ENZOBMLDS0560050
5(S),6(R)-Dihydroxyeicosa-7E,9E,11Z,14Z-tetraenoic acid (DiHETE) ≥99% (durch HPLC)
5(S),6(R)-Dihydroxyeicosa-7E,9E,11Z,14Z-tetraenoic acid (DiHETE)

Synthesis via enzymatic and nonenzymatic hydrolysis of LTA4. Induces contraction of guinea pig lung parenchymal strips. Partial agonist at LTD4 receptors. Potent inhibitor of LTB4 induced neutrophil migration (IC50 = 8nM).


  • Prostanoid receptor ligand


Colorless liquid.

Formel: C₂₀H₃₂O₄
Lagertemperatur: Ultra-Tiefkühlschrank (–80°C)
CAS-Nummer: 82948-88-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c7ed3b41b7e2b3a66f9e590ee49a63bf765b0deb&vid=1279df8a7c4e0f348d315a881528d9d174a9faee&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR