(R)-(-)-2-Amino-5-phosphonopentanoic acid ≥99% (durch HPLC), weißer bis weißgrauer Feststoff
Lieferant: ENZO LIFE SCIENCES
|
Achtung
|
Synonyme:
D-2-Amino-5-phosphonovaleric acid, (R)-2-Amino-5-phosphonopentanoic acid, D-AP5
BMLEA1040005
ENZOBMLEA1040005EA
165
EUR
ENZOBMLEA1040005
(R)-(-)-2-Amino-5-phosphonopentanoic acid ≥99% (durch HPLC), weißer bis weißgrauer Feststoff
(R)-(-)-2-Amino-5-phosphonopentanoic acid
NMDA receptor antagonist.
Active form of AP5. Potent NMDA receptor antagonist.
Soluble in water (acidify for better solubility).
Bitte beachten Store, as supplied, at room temperature for up to 1 year. Store solutions at -20 °C for up to 3 months.
Formel:
C₅H₁₂NO₅P Molecular Weight: 197,127 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00078839 CAS-Nummer: 79055-68-8 |
Spezifikation Testergebnisse
Purity | ≥99% (HPLC) |
Identity | Determined by H-NMR |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...