AMPA ≥98% (durch TLC), weißer bis weißgrauer Feststoff
Lieferant: ENZO LIFE SCIENCES
Synonyme:
3-(3-Hydroxy-5-methylisoxazol-4-yl)-DL-alanine
BMLEA1100005
ENZOBMLEA1100005EA
161
EUR
ENZOBMLEA1100005
AMPA ≥98% (durch TLC), weißer bis weißgrauer Feststoff
AMPA
AMPA receptor ligand
Potent and selective quisqualate analog that interacts with the AMPA subgroup of ionotropic glutamate receptors.
Soluble in water (2 mg/ml, warm) or DMSO (10 mg/ml).
Formel:
C₇H₁₀N₂O₄ Molecular Weight: 186,2 g/mol Lagertemperatur: Gefrierschrank |
CAS-Nummer:
77521-29-0 |
Spezifikation Testergebnisse
Source | Synthetic |
Purity | ≥98% |
Identity | Determined by NMR |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...