Ibotenic acid ≥98% (durch HPLC), weißer bis weißgrauer Feststoff
Lieferant: ENZO LIFE SCIENCES
|
|
Synonyme:
α-Amino-3-hydroxy-5-isoxazoleacetic acid, 2-Amino-2-(3-oxo-2,3-dihydroisoxazol-5-yl)acetic acid
BMLEA1200001
ENZOBMLEA1200001EA
165
EUR
ENZOBMLEA1200001
Ibotenic acid ≥98% (durch HPLC), weißer bis weißgrauer Feststoff
Ibotenic acid
mGluR agonist. Potent glutamate receptor agonist. Displays full agonist activity at mGlu1 and mGlu5.
Soluble in water (20 mg/ml).
Bitte beachten Store, as supplied, at room temperature for up to 1 year. Store solutions at -20 °C for up to 3 months.
Formel:
C₅H₆N₂O₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00069294 CAS-Nummer: 2552-55-8 UN: 3462 ADR: 6.1,III |
|
Spezifikation Testergebnisse
Identity | Determined by H-NMR and MS |
Purity | ≥98% (HPLC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...