(±)11,12-Epoxyeicosa-5Z,8Z,14Z-trienoic acid ≥98%
Lieferant: ENZO LIFE SCIENCES
BMLEE110050
ENZOBMLEE110050EA
86.9
EUR
ENZOBMLEE110050
(±)11,12-Epoxyeicosa-5Z,8Z,14Z-trienoic acid ≥98%
(±)11,12-Epoxyeicosa-5Z,8Z,14Z-trienoic acid
Biosynthesis via liver and renal cytochrome P-450. Induces glucagon release from pancreatic islets. Vasodilator in intestinal microcirculation. Inhibits Na+K+-ATPase. Urinary excretion in humans is increased during pregnancy-induced hypertension.
- Inhibits Na+K+-ATPase
Biosynthesis via liver and renal cytochrome P-450. Induces glucagon release from pancreatic islets. Vasodilator in intestinal microcirculation. Inhibits Na+K+-ATPase. Urinary excretion in humans is increased during pregnancy-induced hypertension.
Lagertemperatur:
Ultra-Tiefkühlschrank (–80°C) |
CAS-Nummer:
123931-40-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...