Aesculetin ≥98%

Lieferant: ENZO LIFE SCIENCES

Synonyme: 6,7-Dihydroxycoumarin, 6,7-Dihydroxy-2H-chromen-2-one

BMLEI1000001
ENZOBMLEI1000001EA 50.6 EUR
ENZOBMLEI1000001
Aesculetin ≥98%
Esculetin

Lipoxygenase inhibitor


Inhibitor of 5-lipoxygenase (ID50=4 µM) and 12-lipoxygenase (ID50=2.5 µM). Does not inhibit cyclooxygenase.

Formel: C₉H₆O₄
Molecular Weight: 178,14 g/mol
MDL: MFCD00006874
CAS-Nummer: 305-01-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3cb776cb27c9b460b418914b9bc54df5d7e23d9&vid=62f79b4d2c29db2585aa162f92de477c9c85d92c&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR