17-Octadecinsäure ≥99%

Lieferant: ENZO LIFE SCIENCES

BMLEI1030010
ENZOBMLEI1030010EA 348 EUR
ENZOBMLEI1030010
17-Octadecinsäure ≥99%
17-Octadecinsäure

Fatty acid omega-hydroxylase inhibitor


Potent suicide inhibitor of LTB4 ω-oxidase (IC50<5µM). Potent and selective inhibitor of fatty acid ω-hydroxylase (IC50<100 nM). Blocks biosynthesis of 20-HETE.

Formel: C₁₈H₃₂O₂
Molecular Weight: 280,45 g/mol
CAS-Nummer: 34450-18-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05e0d37347d0baa11c652c1cca2f4943d283b2a6&vid=ed298a08b65eb5d43002aae338b94da31731be58&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR