Baicalein ≥98% (durch TLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
5,6,7-Trihydroxy-2-phenylchromen-4-on, 5,6,7-Trihydroxyflavon
Lipoxygenase inhibitor
Inhibits cellular Ca2+ uptake and calcium mobilization. Inhibitor of protein tyrosine kinase in leukemia (CEM) cells. Induces cell cycle arrest and apoptosis. Anti-inflammatory compound. Has anti-thrombotic, anti-proliferative and anti-mitogenic effects.Originally reported as a selective inhibitor of 12-lipoxygenase, baicalein has since been shown to inhibit both 12-LO and 15-LO (IC50=0.64µM for 12-LO and 1.6µM for 15-LO). Inhibits leukotriene biosynthesis (at higher concentrations) and release of lysosomal enzymes. Inhibits adjuvant-induced arthritis. Also inhibits prolyl oligopeptidase (POP/PREP)(IC50=36µM).
Formel:
C₁₅H₁₀O₅ Molecular Weight: 270,24 g/mol |
MDL:
MFCD00017459 CAS-Nummer: 491-67-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...