4-Phenylchalcone oxide (synthetisch) ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

BMLEI1080100
ENZOBMLEI1080100EA 429 EUR
ENZOBMLEI1080100
4-Phenylchalcone oxide (synthetisch) ≥98% (durch HPLC)
4-Phenylchalcone oxide

Potent and selective inhibitor of microsomal and cytosolic epoxide hydrolase (IC50=68 nM). Inhibits epoxide hydrolysis of epoxyeicosatrienoic acids.


  • Epoxide hydrolase inhibitor


White to off-white solid.


Soluble in DMSO (25mg/ml)

Formel: C₂₁H₁₆O₂
Lagertemperatur: Gefrierschrank
MDL: MFCD00211155
CAS-Nummer: 82389-34-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43a104c38a063916cc13ee123df15b084efe342c&vid=35a14d29b9ffebb276ce6ac99182befd09a9faa4&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR