A-3 hydrochloride ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: N-(2-Aminoethyl)-5-chloronaphthalene-1-sulphonamide hydrochloride

BMLEI1450010
ENZOBMLEI1450010EA 86.9 EUR
ENZOBMLEI1450010
A-3 hydrochloride ≥98% (durch TLC)
A-3 hydrochloride

Inhibitor of cAMP- and cGMP-dependent protein kinase (PKA, Ki=4.3µM and PKG, Ki=3.8µM), protein kinase C (PKC, Ki=47µM), casein kinase I and II, and myosin light chain kinase (MLCK, Ki=7.4µM).


  • PKA inhibitor


White to off-white solid.


Soluble in water (50mg/ml), DMSO (50mg/ml) or 100% ethanol.

Formel: C₁₂H₁₃ClN₂O₂S·HCl
Molecular Weight: 284.8 · 36.5 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00077311
CAS-Nummer: 78957-85-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41f99fb1cb30d4dd89757cbdff311f5cbf097f76&vid=c25448392b2585a9d74c8ea82e1fbfcad49120a1&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR