H-7 dihydrochloride ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

BMLEI1480010 BMLEI1480050
ENZOBMLEI1480010EA 114 EUR
ENZOBMLEI1480010 ENZOBMLEI1480050
H-7 dihydrochloride ≥98% (durch TLC)
H-7 dihydrochloride

Inhibits PKA (Ki=3.0μM), PKG (Ki=5.8μM), MLCK (Ki=97μM) and PKC (Ki=6μM). Also inhibits myosin light chain kinase. Induces apoptotic DNA fragmentation and cell death. Inhibits telomerase activity in treated NPC-076 cells. Cell permeable.


  • PKC inhibitor


White to light yellow solid.


Soluble in water (50mg/ml), 100% ethanol or DMSO.

Formel: C₁₄H₁₇N₃O₂S·2HCl
Schmelzpunkt: 219…222 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00036961
CAS-Nummer: 108930-17-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5f2af9ad323b668b4907d910e9e6bc88d3e4852&vid=54ddb51a517f236c3ff609281177431a76bc8d8b&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR