1-O-Hexadecyl-2-O-acetyl-sn-glycerol ≥95% (durch TLC)
Lieferant: ENZO LIFE SCIENCES
BMLEI1500005
ENZOBMLEI1500005EA
165
EUR
ENZOBMLEI1500005
1-O-Hexadecyl-2-O-acetyl-sn-glycerol ≥95% (durch TLC)
1-O-Hexadecyl-2-O-acetyl-sn-glycerol
Inhibits protein kinase C activation by diacylglycerols. Stimulates HL-60 cell differentiation. Biosynthetic precursor of PAF via remodeling and retroconversion/de novo routes. Induces delayed aggregation of rabbit platelets. Mitogenic for smooth muscle cells.
- PKC inhibitor
Waxy solid.
Soluble in 100% ethanol or acetone.
Formel:
C₂₁H₄₂O₄ Molecular Weight: 358,6 g/mol Lagertemperatur: Gefrierschrank |
CAS-Nummer:
77133-35-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...