Phloretin ≥98%

Lieferant: ENZO LIFE SCIENCES

BMLEI1540200
ENZOBMLEI1540200EA 89.1 EUR
ENZOBMLEI1540200
Phloretin ≥98%
Phloretin

PKC inhibitor


Flavonoid. Protein kinase C (PKC) inhibitor. PGF2a receptor antagonist. In astrocytes, these receptors are linked to phospholipase C, thus affecting phosphoinositide hydrolysis and intracellular Ca2+ levels. Inhibits myo-inositol uptake and 5’-iodothyronine deiodinase. Prevents TNF-α stimulated upregulation of VCAM-1, ICAM-1 and E-selectin. Enhances adiponectin expression.

Formel: C₁₅H₁₄O₅
Molecular Weight: 274,28 g/mol
MDL: MFCD00002288
CAS-Nummer: 60-82-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ff90a03f79b14180e9effab5feccb72b2b294dd&vid=437fe8e03845f5ecd1e62f70d14e001def50ee3f&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR