3-Methylcyclopentadecanon ≥99% (durch TLC), farbloses Öl
Lieferant: ENZO LIFE SCIENCES
Synonyme:
3-Methylcyclopentadecanone, Muscone
BMLEI1610500
ENZOBMLEI1610500EA
348
EUR
ENZOBMLEI1610500
3-Methylcyclopentadecanon ≥99% (durch TLC), farbloses Öl
3-Methylcyclopentadecanon
Induces rat liver microsomal cytochrome P-450.
Soluble in 100% ethanol (25 mg/ml). Insoluble in water.
Formel:
C₁₆H₃₀O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00211114 CAS-Nummer: 541-91-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...