2-(4-Isobutylphenyl)propansäure ≥99%
Lieferant: ENZO LIFE SCIENCES
Synonyme:
(±)-Ibuprofen, [(±)-2-(4-Isobutylphenyl)-propionic Acid, Ibuprofen
BMLEI1640005
ENZOBMLEI1640005EA
119
EUR
ENZOBMLEI1640005
2-(4-Isobutylphenyl)propansäure ≥99%
2-(4-Isobutylphenyl)propanoic acid
COX inhibitor
Selective cyclooxygenase (COX) inhibitor (IC50=14.9 µM). Inhibits PGH synthase-1 and PGH synthase-2 with comparable potency (ID50=8.9-14 and 7.2-8.2 µM respectively; data are for (S) isomer). Clinically useful NSAID. Inhibits the neutrophil-dependent edema response of rabbit skin to C5a/PGE2.
Formel:
C₁₃H₁₈O₂ Molecular Weight: 206,3 g/mol |
MDL:
MFCD00010393 CAS-Nummer: 15687-27-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...