Aristolochic acid sodium salt ≥97%
Lieferant: ENZO LIFE SCIENCES
BMLEI1760050
ENZOBMLEI1760050EA
110
EUR
ENZOBMLEI1760050
Aristolochic acid sodium salt ≥97%
Aristolochic acid sodium salt
Water soluble form of aristolochic acid.
- Phospholipase inhibitor
Yellow solid.
Soluble in water.
Formel:
C₁₇H₁₀NNaO₇ Molecular Weight: 363.2 g/mol Lagertemperatur: Raumtemperatur |
CAS-Nummer:
10190-99-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...