N-(4-n-Pentylcinnamoyl)anthranilic acid ≥99% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: N-(p-Amylcinnamoyl)anthranilic acid, 2-[3-(4-Pentylphenyl)prop-2-enoylamino]benzoicacid, 2-(3-(4-Pentylphenyl)acrylamido)benzoic acid

BMLEI1780250
ENZOBMLEI1780250EA 348 EUR
ENZOBMLEI1780250
N-(4-n-Pentylcinnamoyl)anthranilic acid ≥99% (durch HPLC)
N-(4-n-Pentylcinnamoyl)anthranilic acid

ACA is a close structural analog of the PLA2 inhibitor, ONO-RS-082 and possesses similar but less potent inhibitory activity. Inhibits TRP channels independently of PLA2, including TRPM2, TRPC6 and TRPM8. It is a useful tool for studying the physiological role of TRP channels.


  • Inhibitor of PLA2 and TRP channels


White solid.


Soluble in DMSO (50 mg/ml) or ethanol (50 mg/ml @ 60°C).

Formel: C₂₁H₂₃NO₃
Molecular Weight: 337,419 g/mol
Schmelzpunkt: 75…77 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00211365
CAS-Nummer: 110683-10-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR