Tyrphostin A25 ≥99%

Lieferant: ENZO LIFE SCIENCES

BMLEI1870005
ENZOBMLEI1870005EA 86.9 EUR
ENZOBMLEI1870005
Tyrphostin A25 ≥99%
Tyrphostin A25

A potent inhibitor of EGF receptor kinase activity (IC50=3 µM).


  • EGFR inhibitor


Yellow-orange solid.


Soluble in 100% ethanol or DMSO.

Formel: C₁₀H₆N₂O₃
Molecular Weight: 202,169 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00133900
CAS-Nummer: 118409-58-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=34c3000ceeb45ab0c9c6134a3cec7a3407219ba0&vid=21db5c02eea7f935eb404600ee086c8cd5046a30&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR