Tyrphostin AG 183 ≥99%

Lieferant: ENZO LIFE SCIENCES

Synonyme: Tyrphostin A51, 3-Amino-2,4-dicyano-5-(3',4',5'-trihydroxyphenyl)penta-2,4-dienonitrile

BMLEI1890005
ENZOBMLEI1890005EA 110 EUR
ENZOBMLEI1890005
Tyrphostin AG 183 ≥99%
Tyrphostin AG 183

A potent inhibitor of EGF receptor kinase activity (IC50=0.8 µM). Inhibition is mixed competitive with ATP and substrate.


  • EGFR inhibitor


Brown solid.


Slightly soluble in water.

Formel: C₁₃H₈N₄O₃
Molecular Weight: 268.2 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00133903
CAS-Nummer: 122520-90-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65db510e4d6837a4e5856e5800388279af7baf4e&vid=4a261b1cc8c82a9be81154c00f082f6036c81b9f&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR