KN-62 ≥98%
Lieferant: ENZO LIFE SCIENCES
Synonyme:
(S)-4-(2-(N-Methylisoquinoline-5-sulphonamido)-3-oxo-3-(4-phenylpiperazin-1-yl)propyl)phenyl isoquinoline-5-sulphonate
BMLEI2300001
ENZOBMLEI2300001EA
181
EUR
ENZOBMLEI2300001
KN-62 ≥98%
KN-62
CaM kinase inhibitor
KN-62 is a potent and selective inhibitor of Ca2+/calmodulin kinase II (Ki=0.9 µM). Causes cells to arrest in S phase. In HEK-293 cells expressing type III adenylyl cyclase (III-AC), 10 µM blocked CaMKII phosphorylation of III-AC and inhibited hormone-stimulated Ca2+ oscillations.
Formel:
C₃₈H₃₅N₅O₆S₂ Molecular Weight: 721,84 g/mol |
MDL:
MFCD00083180 CAS-Nummer: 127191-97-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...