5,6-Dichlor-1-(β-D-ribofuranosyl)-1H-benzimidazol ≥97% (durch TLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
5,6-Dichloro-1-(β-D-ribofuranosyl)benzimidazole, 5,6-Dichlorobenzimidazole Riboside, (2R,3R,4S,5R)-2-(5,6-Dichloro-1H-benzo[d]imidazol-1-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, DRB
DRB is a potent and specific inhibitor of casein kinase II (CKII, IC50~6 µM). It has been used to inhibit RNA polymerase II mediated-transcription which may be dependent on CKII and its interaction with ATF-1.
- Casein kinase inhibitor
White solid.
Soluble in DMSO (20mg/ml), 100% ethanol, and dimethyl formamide (100mM).
Formel:
C₁₂H₁₂Cl₂N₂O₄ Molecular Weight: 319,14 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00036785 CAS-Nummer: 53-85-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...