U-0521
Lieferant: ENZO LIFE SCIENCES
Synonyme:
1-(3,4-Dihydroxyphenyl)-2-methylpropan-1-one
BMLEI2420100
ENZOBMLEI2420100EA
299
EUR
ENZOBMLEI2420100
U-0521
U-0521
Catechol-O-methyltransferase inhibitor
Catechol-O-methyltransferase (COMT) inhibitor.
Formel:
C₁₀H₁₂O₃ Molecular Weight: 180,203 g/mol |
MDL:
MFCD00211220 CAS-Nummer: 5466-89-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...