RG-13022 ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: 3-(3,4-Dimethoxyphenyl)-2-(pyridin-3-yl)acrylonitrile, RG13022, Tyrphostin RG 13022

BMLEI2520005
ENZOBMLEI2520005EA 127 EUR
ENZOBMLEI2520005
RG-13022 ≥98% (durch TLC)
RG-13022

RG-13022 is a non-phenolic tyrphostin analog, which inhibits the EGF receptor (IC50=5 µM) and PDGF receptor tyrosine kinases, and is long-acting. It inhibits EGF-stimulated HER14 cell proliferation (IC50=1 µM) as well as tumor growth in vivo. It inhibits both EGF- and PDGF-stimulated DNA synthesis in human glioma cell lines. RG-13022 is non-phenolic and therefore possesses less antioxidant activity than other tyrphostins.


  • Tyrosine kinase inhibitor


White solid.


Soluble at 25mg/ml in DMSO or 100% ethanol.

Formel: C₁₆H₁₄N₂O₂
Molecular Weight: 266.3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00236444
CAS-Nummer: 136831-48-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a45218e75c27dce918b2f0e2645289a35b163ac9&vid=ae6a3907ab7e2267c51705bc780400780cce541e&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR