Metyrapone ≥99%

Lieferant: ENZO LIFE SCIENCES

Synonyme: 2-Methyl-1,2-di(pyridin-3-yl)propan-1-one

BMLEI2560200
ENZOBMLEI2560200EA 51.7 EUR
ENZOBMLEI2560200
Metyrapone ≥99%
Metyrapone

Inhibits cytochrome P-450-mediated prostaglandin ω/ω-1 hydroxylase activity.


  • Cytochrome P450 inhibitor


White solid.


Soluble in water (25 mg/ml), DMSO (25 mg/ml) or ethanol (25mg/ml).

Formel: C₁₄H₁₄N₂O
Molecular Weight: 226,34 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00006397
CAS-Nummer: 54-36-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dacf5930dc36e922b448f2457dd028c2590d5d0f&vid=31f025445d36f1c82cdb52a7600875399b41c9a6&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR