1-Aminobenzotriazol ≥99%

Lieferant: ENZO LIFE SCIENCES

BMLEI2590100
ENZOBMLEI2590100EA 161 EUR
ENZOBMLEI2590100
1-Aminobenzotriazol ≥99%
1-Aminobenzotriazole

Cytochrome P450 inhibitor


Suicide substrate for cytochrome P-450. Inhibits oxidative drug metabolism in vivo. Blocks omega-oxidation of arachidonic acid but not epoxidation.

Formel: C₆H₆N₄
MDL: MFCD00132902
CAS-Nummer: 1614-12-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6212026c3f3652afc31473fee837a17b7eb3b39&vid=27a79d4256078a5df817839ed3c76177c076b6cc&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR