N-[2-(Cyclohexyloxy)-4-nitrophenyl]methanesulphonamide ≥98% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
N-(2-(Cyclohexyloxy)-4-nitrophenyl)methanesulphonamide, NS-398, N-[2-(Cyclohexyloxy)-4-nitrophenyl]methanesulphonamide
Cyclooxygenase inhibitor. Selectively inhibits PGH synthase-2 (IC50=1 µM) with no inhibition of PGH synthase-1 at concentrations up to 100 µM. Protects neuronal cultures from LPS-induced neurotoxicity.
- COX inhibitor
Cyclooxygenase inhibitor. Selectively inhibits PGH synthase-2 (IC50=1 µM) with no inhibition of PGH synthase-1 at concentrations up to 100 µM. Protects neuronal cultures from LPS-induced neurotoxicity.
Soluble in 100% ethanol (warm; sonication), DMSO (25mg/ml) or dimethyl formamide. Insoluble in water.
Formel:
C₁₃H₁₈N₂O₅S Lagertemperatur: Raumtemperatur |
MDL:
MFCD00882995 CAS-Nummer: 123653-11-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...