7-Ethoxyresorufin ≥98%
Lieferant: ENZO LIFE SCIENCES
Synonyme:
Ethoxyresorufin, 7-Ethoxy-3H-phenoxazin-3-one, Resorufin ethyl ether
BMLEI2620010
ENZOBMLEI2620010EA
253
EUR
ENZOBMLEI2620010
7-Ethoxyresorufin ≥98%
7-Ethoxyresorufin
Cytochrome P450 substrate
Fluorimetric substrate for cytochrome P450 IA1. Inhibits vasorelaxant responses to acetylcholine.
Formel:
C₁₄H₁₁NO₃ Molecular Weight: 241,246 g/mol |
MDL:
MFCD00037661 CAS-Nummer: 5725-91-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...