Gö 6976 ≥95% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: Go6976, 3-(13-Methyl-5-oxo-6,7-dihydro-5H-indolo[2,3-a]pyrrolo[3,4-c]carbazol-12(13H)-yl)propanenitrile

BMLEI2690500
ENZOBMLEI2690500EA 299 EUR
ENZOBMLEI2690500
Gö 6976 ≥95% (durch HPLC)
Gö 6976

PKC inhibitor


Very potent protein kinase C (PKC) inhibitor with high selectivity for the PKC isotypes alpha and beta. Inhibits PKCalpha (IC50=2.3nM) and PKCß1 (IC50=6.2nM) but has no effect on delta, epsilon or gamma isotypes. Potent antagonist of HIV-1 induction.

Formel: C₂₄H₁₈N₄O
Molecular Weight: 378,425 g/mol
MDL: MFCD00236434
CAS-Nummer: 136194-77-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a18e8f91c383726d13a1efc7930b811f53dcca91&vid=1bcab7e7f9c02c1082824272991ba2fa06defc38&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR