BAY 11-7085 ≥99%

Lieferant: ENZO LIFE SCIENCES

BMLEI2790010 BMLEI2790050
ENZOBMLEI2790010EA 114 EUR
ENZOBMLEI2790010 ENZOBMLEI2790050
BAY 11-7085 ≥99%
BAY 11-7085

NF-kappaB inhibitor


An irreversible inhibitor of cytokine-inducible IkappaBalpha phosphorylation (IC50=10 µM). In intact HUVECs it inhibits TNFalpha-induced phosphorylation and degradation of IkappaBalpha without affecting constitutive phosphorylation. Inhibition of IkappaBalpha phosphorylation and degradation results in the inactivation of NF-kappaB and NF-kappaB-dependent expression of endothelial-leukocyte adhesion molecules, E-selectin, VCAM-1 and ICAM-1. BAY 11-7085 displayed potent anti-inflammatory activity in the rat carrageenan paw edema assay and adjuvant arthritis model.

Formel: C₁₃H₁₅NO₂S
MDL: MFCD01862602
CAS-Nummer: 196309-76-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff56d563ae3470e0b7e1767733a3ed0da4ed907c&vid=ed694ccdeb6570d3a7fc5a78ec8597929916c41f&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR