(S)-(+)-Ketoprofen ≥98,5% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: (S)-2-(3-Benzoylphenyl)propanoic acid, S-(+)-Ketoprofen

BMLEI2880001
ENZOBMLEI2880001EA 236 EUR
ENZOBMLEI2880001
(S)-(+)-Ketoprofen ≥98,5% (durch HPLC)
(S)-(+)-Ketoprofen

Active enantiomer of ketoprofen. Inhibits cyclooxygenase.


  • COX inhibitor


White solid.


Soluble in 100% ethanol (25mg/ml).

Formel: C₁₆H₁₄O₃
Molecular Weight: 254,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00673316
CAS-Nummer: 22161-81-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff5aa77cf6bc8b9754242f5f665ea6d4c9002e60&vid=1652ee4edfe904b7eee19357c6aa3f47eb214c8f&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR