(R)-Ibuprofen ≥98%

Lieferant: ENZO LIFE SCIENCES

Synonyme: (R)-2-(4-Isobutylphenyl)propanoic acid, (R)-(-)-Ibuprofen

BMLEI2900200
ENZOBMLEI2900200EA 161 EUR
ENZOBMLEI2900200
(R)-Ibuprofen ≥98%
(R)-Ibuprofen

Less active enantiomer of ibuprofen.


  • COX inhibitor


White crystalline powder.

Formel: C₁₃H₁₈O₂
Molecular Weight: 206,3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00069290
CAS-Nummer: 51146-57-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ad3e7569efc9f37dbecd972d73031ca25724231&vid=017f1e5d311ce6d45dfbc9fc8fbcfe3532b59f5d&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR