Meloxicam ≥98%

Lieferant: ENZO LIFE SCIENCES

Synonyme: 4-Hydroxy-2-methyl-N-(5-methylthiazol-2-yl)-2H-benzo[e][1,2]thiazine-3-carboxamide 1,1-dioxide, 4-Hydroxy-2-methyl-N-(5-methyl-2-thiazolyl)-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide

BMLEI2920100
ENZOBMLEI2920100EA 180 EUR
ENZOBMLEI2920100
Meloxicam ≥98%
Meloxicam

COX inhibitor


Selective COX-2 inhibitor. Active in vivo. IC50 (human chondrocytes) COX-1 = 36.6µM, COX-2 = 4.7µM

Formel: C₁₄H₁₃N₃O₄S₂
Molecular Weight: 351,40 g/mol
MDL: MFCD00868752
CAS-Nummer: 71125-38-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ad0234d3d31a78abea889773bdeae9dd748edfc&vid=7f5e452a8cb8f8523e55aa6930735e16fc26341b&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR