SB202190 ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: 4-(4-(4-Fluorophenyl)-5-(pyridin-4-yl)-1H-imidazol-2-yl)phenol, SB 202190

BMLEI2940001
ENZOBMLEI2940001EA 165 EUR
ENZOBMLEI2940001
SB202190 ≥98% (durch TLC)
SB202190

Cell permeable, potent and selective p38 MAP kinase inhibitor (IC50=30 nM). Inhibits p38α and ß, but not γ and δ isoforms. Does not inhibit ERK2 or other members of the MAP kinase family or their upstream activators. A useful tool for dissecting the role of p38 in signaling pathways. Induces apoptosis.


  • p38 inhibitor


Pale yellow solid.


Soluble in methanol, DMSO or acetone; slightly soluble in ethyl acetate.

Formel: C₂₀H₁₄FN₃O
Lagertemperatur: Gefrierschrank
MDL: MFCD00941964
CAS-Nummer: 152121-30-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b15ae68089ba997efbf2cf1ee4f511373b90ec67&vid=e40fb40b3942ce5ed57180280cab601df546422c&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR