Gö 6983 ≥95% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: 3-(1-(3-(Dimethylamino)propyl)-5-methoxy-1H-indol-3-yl)-4-(1H-indol-3-yl)-1H-pyrrole-2,5-dione, 3-[1-[3-(dimethylamino)propyl]-5-methoxy-1H-indol-3-yl]-4-(1H-indol-3-yl)-1H-pyrrole-2,5-dione, Go 6983

BMLEI2960500
ENZOBMLEI2960500EA 181 EUR
ENZOBMLEI2960500
Gö 6983 ≥95% (durch HPLC)
Gö 6983

PKC inhibitor. Has been shown to selectively inhibit several PKC isoenzymes (IC50 = 7nM for PKCα and PKCβ; 6nM for PKCγ; 10nM for PKCδ; 60nM for PKCζ). The compound does not effectively inhibit PKCµ (IC50=20 µM) and therefore can be used to differentiate PKCµ from other isoforms.


  • PKC inhibitor


Bright red solid.


Soluble in DMSO.

Formel: C₂₆H₂₆N₄O₃
Molecular Weight: 442,519 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD04040031
CAS-Nummer: 133053-19-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c22eb1d3198c5e4ab2778b0f6e39b147be8c1d4&vid=d8c026da4ab939ef392284e7b3165a41a5b4da5a&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR