1,9-Pyrazoloanthrone ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: JNK Inhibitor II, Anthra[1,9-cd]pyrazol-6(2H)-one, Dibenzo[cd,g]indazol-6(2H)-one, SP600125, SP 600125

BMLEI3050010 BMLEI3050050
ENZOBMLEI3050010EA 84.7 EUR
ENZOBMLEI3050010 ENZOBMLEI3050050
1,9-Pyrazoloanthrone ≥98% (durch TLC)
1,9-Pyrazoloanthrone

Potent, cell permeable, selective, and reversible inhibitor of c-Jun N-terminal kinase (JNK) (IC50=40nM for JNK-1 and JNK-2 and 90nM for JNK-3). The inhibition is competitive with respect to ATP. Exhibits over 300-fold greater selectivity for JNK as compared to ERK1 and p38. Inhibits the phosphorylation of c-Jun and blocks the expression of IL-2, IFN-γ, TNF-α, and COX-2. Blocks IL-1-induced accumulation of phospho-Jun, induction of c-Jun transcription and anti CD3-induced apoptosis of CD4+ CD8+ thymocytes.


  • Jnk inhibitor


Yellow to brown solid.


Soluble in DMSO or methanol.

Formel: C₁₄H₈N₂O
Molecular Weight: 220.231 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00022289
CAS-Nummer: 129-56-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=977c6c93e80fdde02c589cbf32b9649fb1ab6643&vid=be08983f5fe9630be7839ab5cc4e38ef6c5ab7e5&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR