GW5074 ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: 5-Iodo-3-[(3,5-dibromo-4-hydroxyphenyl)methylene]-2-indolinone, 3-(3,5-dibromo-4-hydroxybenzylidene)-5-iodoindolin-2-one, GW 5074

BMLEI3070005 BMLEI3070025
ENZOBMLEI3070005EA 121 EUR
ENZOBMLEI3070005 ENZOBMLEI3070025
GW5074 ≥98% (durch TLC)
GW5074

Potent and selective cell permeable inhibitor of cRAF1 kinase (IC50 = 9 nM) with 100-fold selectivity over CDK1, CDK2, c-src, ERK2, MEK, p38, Tie2, VEGFR2 and c-fm. A useful tool for assessing the involvement of the Ras/Raf-1/ERK pathway in various signaling pathways. Displays neuroprotective effects in vivo through a MEK-ERK and Akt-independent mechanism.


  • Raf inhibitor


Yellow to orange solid.


Soluble in DMSO (>25 mg/ml).

Formel: C₁₅H₈Br₂INO₂
Molecular Weight: 520.94 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD09971042
CAS-Nummer: 220904-83-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b4960ac47f4ec93b85c2fd34720ae49e04ce92af&vid=63f05e2fd89a2a064b9d0a638d1069cec3b326a4&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR