Kenpaullone ≥98%

Lieferant: ENZO LIFE SCIENCES

Synonyme: 9-Bromo-7,12-dihydroindolo[3,2-d][1]benzazepin-6(5H)-one, 9-Bromo-7,12-dihydrobenzo[2,3]azepino[4,5-b]indol-6(5H)-one

BMLEI3100005
ENZOBMLEI3100005EA 299 EUR
ENZOBMLEI3100005
Kenpaullone ≥98%
Kenpaullone

Inhibitor of CDK and GSK


Potent inhibitor of CDK1/cyclin B (IC50=400nM). Also inhibits CDK2/cyclin A (IC50=680nM) , CDK5 (IC50=850nM) and with much less effect other kinases. More recently, kenpaullone has been found to be a useful GSK-3beta inhibitor (IC50=23nM).

Formel: C₁₆H₁₁BrN₂O
Molecular Weight: 327,18 g/mol
MDL: MFCD02683595
CAS-Nummer: 142273-20-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=610466e6c250f757dd37bf34cd276c4dd8cc375a&vid=f3192db5fcd0942acccfb3c9cd3cb34c5b921fb0&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR