Sodium 4-phenylbutyrate ≥98% (by UHPLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
Sodium 4-phenylbutanoate, 4-Phenylbutyric acid sodium salt
Weak histone deacetylase (HDAC) inhibitor. Induces differentiation, growth arrest and apoptosis in a number of cell lines. Protects against cerebral ischemic injury and displays neuroprotective effect in a mouse model of Huntington’s disease. Anti-neoplastic agent and transcriptional regulator. Also acts as an inducer of tumor cytostasis and differentiation. Shows chemical chaperone activity.
- HDAC inhibitor
White to yellowish solid.
Soluble in DMSO or water (25mg/ml).
Formel:
C₁₀H₁₁O₂·Na Molecular Weight: 186,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00800247 CAS-Nummer: 1716-12-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...