2,3-Diphenylquinoxaline-6-carboxylic acid ≥98% (durch TLC)
Lieferant: ENZO LIFE SCIENCES
BMLEI3360010
ENZOBMLEI3360010EA
86.9
EUR
ENZOBMLEI3360010
2,3-Diphenylquinoxaline-6-carboxylic acid ≥98% (durch TLC)
2,3-Diphenylquinoxaline-6-carboxylic acid
Inhibits the translocation of Akt.
- Akt inhibitor
Light green solid.
Soluble in DMSO (25 mg/ml).
Formel:
C₂₁H₁₄N₂O₂ Molecular Weight: 326,36 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00555042 CAS-Nummer: 32387-96-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...