ZM-306416 ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: N-(4-Chloro-2-fluorophenyl)-6,7-dimethoxyquinazolin-4-amine, CB-676475

BMLEI3530001
ENZOBMLEI3530001EA 89.1 EUR
ENZOBMLEI3530001
ZM-306416 ≥98% (durch TLC)
ZM-306416

VEGF receptor tyrosine kinase inhibitor. Inhibits KDR (IC50=100nM) and Flt (IC50=2µM) tyrosine kinases.


  • VEGFR inhibitor


Off-white solid.


Soluble in DMSO (3 mg/ml) or ethanol (1 mg/ml).

Formel: C₁₆H₁₃ClFN₃O₂
Molecular Weight: 333,747 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02683969
CAS-Nummer: 690206-97-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29ed8d2216a35b47b23bda6d5c43e3655653a6ad&vid=37ae32ece5438fb30ff88cadfadf616d50306cdb&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR