PD 98059 ≥95% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
PD98059, 2'-Amino-3'-methoxyflavone, 2-(2-Amino-3-methoxyphenyl)chromone, PD 98059, 2-(2-Amino-3-methoxyphenyl)-4H-1-benzopyran-4-one, 2-(2-Amino-3-methoxyphenyl)-4H-chromen-4-one
PD-98059 is a potent and selective inhibitor of MAP kinase kinase (MEK). It selectively blocks the activation of MEK thereby inhibiting the phosphorylation and the activation of MAP kinase. PD-98059 is cell permeable. In PC12 pheochromocytoma cells, it completely blocked the increase in MAP kinase activity produced by NGF, IC50=2µM. Enhances self-renewal of embryonic stem cells PD-98059 is an invaluable tool to help elucidate the role of the MAPK cascade in a variety of biological systems. Inhibits cell growth and proliferation in acute myelogenous leukemia cell lines by causing cell arrest.
- MEK inhibitor
Pale yellow solid
Soluble in DMSO (6.5mg/ml); 25mg/ml in anhydrous DMSO), methanol, dichlormethane and 100% ethanol (0.6mg/ml).
Formel:
C₁₆H₁₃NO₃ Molecular Weight: 267,284 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00671789 CAS-Nummer: 167869-21-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...