Geldanamycin 98 TLC_ASSAY_METHOD (conjugated to FITC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: (4E,6Z,8S,9S,10E,12S,13R,14S,16R)-13-Hydroxy-8,14,19-trimethoxy-4,10,12,16-tetramethyl-3,20,22-trioxo-2-azabicyclo[16.3.1]docosa-1(21),4,6,10,18-pentaen-9-yl carbamate

BMLEI3610001
ENZOBMLEI3610001EA 305 EUR
ENZOBMLEI3610001
Geldanamycin 98 TLC_ASSAY_METHOD (conjugated to FITC)
Geldanamycin

A novel geldanamycin fluorescent probe that may be used in a fluorescence polarization assay for HSP90 inhibitors. May also be used for detection of cell surface HSP90.


  • HSP90 inhibitor

Formel: C₂₉H₄₀N₂O₉
Molecular Weight: 560.64 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00274570
CAS-Nummer: 30562-34-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9f2eeda2fade38200833570477fa8aea3b9351b&vid=ca3bc4812d992c8dc72883d4d186a936443924df&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR